Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN75, 15 mm
Product Code. 17518765 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SN75 Sterilizer
Unit Size:
Each
Product Code. 17518765
Brand: Memmert™ SN75B02072
Only null left
Add to basket
Add to basket
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) SN75 Sterilizer
Product Type Drip Tray
Height (Metric) Tray Rim: 15 mm
Material Stainless Steel