Learn More
Bochem™ Stainless Steel Spatulas
Click to view available options
Length (Metric):
165 mm
250 mm
275 mm
365 mm
415 mm
Length (Metric) Blade:
75 mm
150 mm
165 mm
250 mm
300 mm
Unit Size:
Each
Pack of 10
Description
- Wood handle
- Manufactured from stainless steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Spatula |
| Length (Metric) Blade | 300 mm |
| Width (Metric) | 42 mm |
| Width (Metric) Blade | 42 mm |
| Blade Material | Stainless Steel |
| Diameter (Metric) Outer | 1mm |
| Dimensions (L x W) | 415 x 42 mm |
| Handle Material | Wood |
| Material | Stainless Steel |
| Show More |