Learn More
Agencinox™ Stainless Steel Sterilization Boxes
Product Code. 11703944
Click to view available options
Height (Metric):
40 mm
50 mm
60 mm
70 mm
100 mm
Unit Size:
Each
Product Code. 11703944
Brand: Agencinox™ 5052
Use these boxes for a variety of general laboratory applications. Agencinox™ Stainless Steel Sterilization Boxes keep instruments safe and secure during sterilization process.
  • Stainless steel instrument boxes
  • Available in a variety of sizes
Specifications
Material Stainless Steel
Shape Rectangular
Product Type Sterilization Box
Length (Metric) 300 mm
Width (Metric) 150 mm
Height (Metric) 70 mm