missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Stainless Steel Storage Racks
Brand: Cole-Parmer™ 36850-43
Additional Details : Weight : 1.36000kg
Specifications
Autoclavable | |
Yes | |
1 Each |
400 mL | |
For 12 Bags 400mL |