missing translation for 'onlineSavingsMsg'
Learn More

Arex Professional Equipment™ Stainless Steel Table with Bottom Shelf, 60 x 120 x 85 mm

Constructed completely of stainless steel with level adjustable legs.

Brand:  Arex Professional Equipment™ CTR612S

Additional Details : Weight : 32.00000kg

Product Code. 11894290

  • 1142.10€ / Each
Estimated Shipment:
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Stainless steel construction throughout.
  • Adjustable legs.
  • Convenient bottom shelf.
Specifications

Specifications

Bottom Shelf
85 mm
120 mm
Stainless Steel
60 x 120 x 85 mm
60 mm
Stainless Steel
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions