Learn More
Arex Professional Equipment™ Stainless Steel Table with Bottom Shelf, 70 x 180 x 85 mm
Constructed completely of stainless steel with level adjustable legs.
Brand:  Arex Professional Equipment™ CTR718S
Additional Details : Weight : 40.00000kg
Product Code. 11894310
  • POA / Each
Estimated Shipment: 19-08-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Stainless steel construction throughout.
  • Adjustable legs.
  • Convenient bottom shelf.
Specifications
Specifications
Bottom Shelf
85 mm
180 mm
Stainless Steel
70 x 180 x 85 mm
70 mm