Learn More
Knick Elektronische™ Stainless Steel Temperature Sensor
Product Code. 10199650
Click to view available options
For Use With (Application):
For pH meter
Unit Size:
Each
Récupel-bijdrage uitgesloten
Product Code. 10199650
Brand: Knick Elektronische™ ZU0156
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
PT 1000 temperature probe for use with SE 104 N pH Sensor.
  • PT 1000 temperature probe
  • For use with SE 104 N pH Sensor
Specifications
Type Temperature Sensor
For Use With (Application) For pH meter
Material Stainless Steel