Learn More
Grant Instruments™ Stainless Steel Test Tube Rack
Product Code. p-8011789
Click to view available options
For Use With (Equipment):
0.5mm dia. Tubes
1.5mm dia. Tubes
10mm dia. Tubes
13mm dia. Tubes
16mm dia. Tubes
19mm dia. Tubes
25mm dia. Tubes
30mm dia. Tubes
Unit Size:
Each
Product Code. 15385842
Brand: Grant Instruments™ SRSE
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Fits a variety of test tube sizes from 10 to 30mm diameter
  • For use with LSB Aqua Pro
  • Available in 8 sizes to hold up to 48 SR test tubes
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 119 mL
For Use With (Equipment) 0.5mm dia. Tubes
Material Stainless Steel