Learn More
JP Selecta™ Stainless Steel Tripod Support Stand
Product Code. 12012365
Click to view available options
Diameter (Metric) Outer:
8 mm
10 mm
12 mm
Height (Metric):
500 mm
600 mm
700 mm
Weight (Metric):
1.4 kg
1.8 kg
2.5 kg
Unit Size:
Each
Product Code. 12012365
Brand: JP Selecta™ 6022710
Only null left
Add to basket
Add to basket
JP Selecta™ Tripod Support Rods is suitable for laboratory purposes to support tripod.
  • Made of AISI 304 stainless steel
Specifications
Material Stainless Steel
Height (Metric) 600 mm
Diameter (Metric) Outer 10 mm
Product Type Tripod support rod
Quantity 1 Ea.
Weight (Metric) 1.8 kg