Learn More
RUBIS Long Tail Tweezers
Long Tail Tweezers - Inox Steel, Serrated Handle
33.05€ - 44.06€
Specifications
Tip Style Straight, Round, Blunt, Serrated
Material Inox Steel
Product Type Long Tail Tweezers
Quantity (No. per Case) Each
Type Tweezers
View More Specs
Products 3
Product Code Brand Length (Metric) Length (English) Price Quantity & Availability  
Product Code Brand Length (Metric) Length (English) Price Quantity & Availability  
11719165
View Documents Promotion Details
RUBIS
232/8
215 mm 8.46 in.
33.05€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-04-2024
to see stock.
Add to basket
11729165
View Documents Promotion Details
RUBIS
232/10
255 mm 10.04 in.
39.26€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-06-2024
to see stock.
Add to basket
11739165
View Documents Promotion Details
RUBIS
232/12
300 mm 11.81 in.
44.06€
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
Description
Description
  • Long Tail Tweezers
  • 100% Made in Switzerland
  • Anti-acid/Antimagnetic Stainless Steel
  • Serrated handles
  • Serrated tips for better grip
  • Blunt tips
  • Suitable for cryogenics applications
  • Strong
Specifications
Specifications
Straight, Round, Blunt, Serrated
Long Tail Tweezers
Tweezers
Inox Steel