Learn More
RUBIS Wafer Handling Tweezers
Wafer Handling Tweezers - Antimagnetic/Anti-acid SS, Serrated Handle
27.84€ - 41.86€
Specifications
Material Anti-acid, Anti-magnetic Stainless Steel
Product Type Wafer Handling Tweezers
View More Specs
Products 3
Product Code Brand Length (Metric) Design Price Quantity & Availability  
Product Code Brand Length (Metric) Design Price Quantity & Availability  
11703426
View Documents Promotion Details
RUBIS
39S4
130 mm Top Lip, Stepped Perforated Bottom Paddle
35.55€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
11793416
View Documents Promotion Details
RUBIS
ARG
130 mm Top Lip, Stepped Bottom Paddle
41.86€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
11723276
View Documents Promotion Details
RUBIS
2LB/4
125 mm Top Lip, Stepped Bottom Paddle
27.84€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • Wafer Handling Tweezers
  • High Precision Tweezers
  • 100% Made in Switzerland
  • Anti-acid/Antimagnetic Stainless Steel
  • Perfect alignment
  • Semiconductor industry
  • Very Reliable
Specifications
Specifications
Anti-acid, Anti-magnetic Stainless Steel
Wafer Handling Tweezers