Learn More
Bochem™ Stainless Steel Z Shape Test Tube Stands
Click to view available options
Dimensions (L x W):
50 x 85 mm
50 x 110 mm
50 x 220 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Test Tube Rack |
| Holds | Tubes (17 mm) |
| Material | Stainless Steel (18/10) |
| Positions | 50 |
| Dimensions (L x W) | 50 x 220 mm |
| Diameter (Metric) Opening | 17 mm |