Learn More
Fisherbrand™ Retort Stand Assembly for Cyclone Mixers
Ideal for customers who require a permanent fixing for test tubes during long term or frequent tube mixing.
Brand:  Fisherbrand™
Additional Details : Weight : 0.50000kg
Product Code. 10162224
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Ideal for customers who require a permanent fixing for test tubes during long term or frequent tube mixing
  • It can also be used to reduce 'finger or hand' fatigue in orbital mixing
  • Also on open tubes during mixing it offers a safer option capping the tube 
  • The accessory kit comprises of retort rod and clamp arm
  • The retort rod conveniently screws into a fixing point on the the Cyclone Mixer case
  • This accessory clamps tubes or vials onto the rubber cup for mixing
Specifications
Specifications
Stand Assembly
Cyclone Vortex Mixer (CM-1)