Learn More
Standard Buffer Solution, SI Analytics™
Product Code. 10721211 Shop All SI Analytics Products
Click to view available options
Quantity:
250 mL
60 Ampules
Unit Size:
250mL
60 ampoules
Product Code. 10721211
Brand: SI Analytics 285138016
SI Analytics™ Standard Buffer Solution pH 6.87 resists changes in pH when small quantities of an acid are added to it.
  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, disodium hydrogen phospate, potassium dihydrogen phosphate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 100°C
Color Colorless
Density 1g/cm3
pH 6.87
Physical Form Liquid
Quantity 250 mL
Formulation Water, disodium hydrogen phospate, potassium dihydrogen phosphate
Odor Odorless
Packaging DURAN™ Bottle
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water