Learn More
Standard Buffer Solution pH 7, SI Analytics™
Product Code. p-8014351 Shop All SI Analytics Products
Click to view available options
Quantity:
250 mL
60 Ampules
Unit Size:
250mL
Each
Product Code. 10131311
Brand: SI Analytics 285138221
SI Analytics™ Standard Buffer Solution pH 7 resists changes in pH when small quantities of an acid or an alkali are added to it.
  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, disodium hydrogenphosphate, citric acid
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 101°C
Color Colorless
Density 1.01g/cm3
pH 7
Physical Form Liquid
Quantity 60 Ampules
Formulation Water, disodium hydrogenphosphate, citric acid
Odor Odroless
Packaging FIOLAX™ Ampules
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water