missing translation for 'onlineSavingsMsg'
Learn More

Standard Buffer Solution, SI Analytics™

Catalog Number p-8014352
Click to view available options
Quantity:
250 mL
60 Ampules
This item is not returnable. View return policy
This item is not returnable. View return policy

SI Analytics™ Standard Buffer Solution pH 9.18 resists changes in pH when small quantities of an alkali are added to it.

  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, disodium tetraborate decahydrate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
Boiling Point 100°C
Color Colorless
Density 1g/cm3
pH 9.18
Physical Form Liquid
Quantity 60 Ampules
Formulation Water, disodium tetraborate decahydrate
Odor Odorless
Packaging FIOLAX™ Ampules
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'