Learn More
Sartorius™ Standard Flexel™ 3D bags with silicone tubes
Brand:  Sartorius™ FXB103364
Additional Details : Weight : 4.00000kg
Product Code. 12939330
  • 3079.30€ / Pack of 5
Estimated Shipment: 30-07-2024
to see stock.
Specifications
200 L
Bags are designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products
5/Pk.
ISO11137
3 Ports
Gamma Irradiated