Learn More
BINDER™ Standard Rack
Product Code. 30238349 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB 65
KB PRO/KBF/KBF PRO 1600
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 130, KBF/KBF PRO 130
KB/KB PRO 720, KBF/KBF PRO 720
KB/KB PRO/KBF/KBF PRO 260/470
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
30238349 KB 65 Each
30238365 KB/KB PRO 130, KBF/KBF PRO 130 Each
30238271 KB/KB PRO/KBF/KBF PRO 260/470 Each
30238345 KB/KB PRO 720, KBF/KBF PRO 720 Each
30238295 KB/KB PRO 1060, KBF/KBF PRO 1060 Each
30238360 KB PRO/KBF/KBF PRO 1600 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30238349 Supplier BINDER™ Supplier No. 80122388
Only null left
Add to basket
Add to basket
Shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
  • Shelf rack made of stainless steel
  • Includes set of U-rails
Specifications
For Use With (Equipment) KB 65
Material Stainless Steel
Type Rack