Learn More
Ismatec Standard Tubing, Tygon LMT-55
Inexpensive, all-around tubing for general laboratory applications.
Brand:  Ismatec SC0376T
Additional Details : Poids : 0.80000kg
 View more versions of this product
Product Code. 15383549
  • 118.00€ / 15m
Expédition estimée
22-10-2020
Add to basket
Description
Description
  • Transparent.
  • Resistant to almost all inorganic chemicals.
  • Smooth polished inner wall.
  • Low gas permeability.
  • Non-aging and non-oxidizing.
Specifications
Specifications
Transparent
Tygon LMT-55
15 m
−50°C
Acids: Good, Alkaline solutions: Good, Solvents: Not rec., Pressure: Fair, Vacuum: Good, Viscous media: Excellent, Sterile media: Limited
8.0 mm
11.2 mm
General laboratory applications.
1 roll
I.D. 8.0 mm, O.D. 11.2 mm