Learn More
BOLA Star Head Magnetic Stirring Bars
Click to view available options
Diameter (Metric):
35 mm
40 mm
58 mm
Unit Size:
Each
Description
PTFE-encapsulated magnetic core (Alnico 5), universal chemical resistance.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | White |
| Height (Metric) | 12 mm |
| Coating | PTFE |
| Diameter (Metric) | 35 mm |
| Shape | Star |