Learn More
Starch Indicator Solution, Chem Lab™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
500mL
Each
Specifications
Specifications
| Melting Point | 0°C |
| Solubility Information | Soluble in water |
| Odor | Odorless |
| Color | Colorless |
| pH | ±7 |
| Quantity | 100 mL |
| Physical Form | Liquid |