Learn More
Abeba™ Static Control 1275 Shoes
ESD dissipative and Sanitized™ treated.
Brand:  Abeba™ 1275/44
Additional Details : Weight : 1.25000kg
 View more versions of this product
Product Code. 15627217
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • CE EN ISO 20345:2011 S1 SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Composite toe cap
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Replaceable acc Wave insole (article no. 3553)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Double velcro fastening
  • Reflective material
  • Leather with scratch-resistant carbon coating
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRA
Leather
Slip-resistant
1275
Black
44
Rubber
Composite