Learn More
Abeba™ Static Control 31362 Shoes
ESD dissipative and Sanitized™ treated.
Brand:  Abeba™ 31362/37
Additional Details : Weight : 1.40000kg
 View more versions of this product
Product Code. 15698807
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather
  • Black
  • Composite toe cap
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Replaceable acc Wave insole (article no. 3553)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRA
Leather
Slip-resistant
31362
Black
37
Rubber
Composite