Learn More
Abeba™ Static Control 31362 Shoes
ESD dissipative and Sanitized™ treated.
Brand: Abeba™ 31362/37
Additional Details : Weight : 1.40000kg
Description
- CE EN ISO 20345:2011 S2 SRA
- Smooth leather
- Black
- Composite toe cap
- PU midsole
- Slip resistance special rubber outsole sole
- Replaceable acc Wave insole (article no. 3553)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Leather | |
Slip-resistant | |
31362 |
Black | |
37 | |
Rubber | |
Composite |