Learn More
Abeba™ Static Control 31366 Shoes
ESD dissipative and Sanitized™ treated.
Brand: Abeba™ 31366/45
Additional Details : Weight : 1.30000kg
Description
- CE EN ISO 20345:2011 S1 SRA
- Velours
- Anthracite
- Composite toe cap
- PU midsole
- Slip resistance special rubber outsole sole
- Replaceable acc Wave insole (article no. 3553)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Specifications
CE EN ISO 20345:2011 S1 SRA | |
Velour | |
Slip-resistant | |
31366 |
Anthracite | |
45 | |
Rubber | |
Composite |