Learn More
Abeba™ Static Control 31378 Shoes
ESD dissipative and Sanitized™ treated.
Brand:  Abeba™ 31378/43
Additional Details : Weight : 1.25000kg
 View more versions of this product
Product Code. 15678887
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S1 SRA
  • Velours with breathable Air-Mesh textile inlays
  • Anthracite
  • Composite toe cap
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Replaceable acc Wave insole (article no. 3553)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Adjustable instep area
  • Double velcro fastening
  • Reflective material
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRA
Velour
Slip-resistant
31378
Anthracite
43
Rubber
Composite