Learn More
Rogo Sampaic™ Galvanized Steel Opened Rings
Product Code. p-8009978
Click to view available options
Diameter (Metric) Outer:
60 mm
80 mm
100 mm
140 mm
Unit Size:
Each
Product Code. 11752654
Brand: Rogo Sampaic™ FCR501
Only null left
Add to basket
Add to basket
Provides for ease of connection on ring stands. Rogo Sampaic™ Galvanized Steel Opened Rings are an economical and durable choice.
  • Available in a range of diameters
  • Helps support a variety of glassware and other chemistry equipment
Specifications
Material Steel
Diameter (Metric) Outer 60 mm
Product Type Opened Ring
Quantity 1 Ea.