missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ STELLAR Safety Spectacles

Product Code. 12684913
Click to view available options
:
Product Type: Safety Glasses
This item is not returnable. View return policy

Product Code. 12684913

Brand: Honeywell Safety Products™ 1005564

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Stylish, spectacle-type impact protection in a lightweight polycarbonate lens. Honeywell Safety Products™ STELLAR Safety Spectacles offer protection and the comfort of quality prescription eyewear.

  • General use and protection against impact (45m/s).
  • Suitable for mechanic work: grinding, turning, milling.
  • Polycarbonate lenses offer 99.9% UV protection up to 380nm.
  • Stylish safety spectacle offering the comfort of quality prescription eyewear.
  • Quality metal frame design.
  • Hardcoat for better abrasion resistance.
  • Adjustable, breakaway Flexicord retainer.
  • Adjustable hypoallergenic nose pads and metal temples with 3D spatula tips.
  • Excellent fit.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Safety glasses
No. per Pack 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.