missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Cardiomyocyte Differentiation Kit

Product Code. 15735778
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15735778

Brand: STEMCELL Technologies 05010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Media for differentiation of human PSCs to cardiomyocytes and long-term maintenance of human PSC-derived cardiomyocytes

  • STEMdiff™ Cardiomyocyte Differentiation Kit (Catalog #05010) includes a medium for differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells (human pluripotent stem cells [hPSCs]) into cardiomyocytes (cardiac troponin T-positive [cTnT+]), as well as a medium for maintenance of hPSC-derived cardiomyocytes
  • This kit can be used to generate cardiomyocytes derived from a clump culture of hPSCs maintained in mTeSR™1 (Catalog #85850), mTeSR™ Plus (Catalog #100-0276), or TeSR™-E8™ (Catalog #05990)
  • Greater than 80% of these cells will be cTnT+
  • An average of 1 x 10^6 cells can be harvested from a single well of a 12-well plate
  • STEMdiff™ Cardiomyocyte Maintenance Kit (Catalog #05020) comprises the maintenance basal medium and supplement; it can be used for long-term maintenance of hPSC-derived cardiomyocytes for one month or longer
  • These cardiomyocytes can be used in various downstream applications and analyses

Specifications

Cell Type Cardiomyocytes, PSC-Derived
For Use With (Application) Cell Culture, Differentiation, Maintenance
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.