missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Pancreatic Progenitor Kit

Product Code. 17178251
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17178251

Brand: STEMCELL Technologies 05120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Serum-free, defined medium for the differentiation of human ES and iPS cells to pancreatic progenitor cells

  • STEMdiff™ Pancreatic Progenitor Kit is a serum-free medium that supports efficient and reproducible generation of pancreatic progenitor cells from human pluripotent stem cells (hPSCs)
  • Cells are differentiated through four stages: 1) definitive endoderm, 2) primitive gut tube, 3) posterior foregut endoderm, and 4) pancreatic progenitor cells
  • The differentiated cells express key markers of pancreatic progenitor cells, including PDX-1, NKX6.1 and SOX9 and up-regulation of insulin and glucagon
  • The resulting pancreatic progenitor cells can be further differentiated to insulin-producing beta cells or other endocrine and exocrine pancreatic cell fates
  • STEMdiff™ Pancreatic Progenitor Kit has been optimized for differentiation of cells maintained in mTeSR™1 (Catalog #85850)

Specifications

Cell Type Pancreatic Cells, Pluripotent Stem Cells
For Use With (Application) Cell Culture, Differentiation
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.