missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies StemSpan™ SFEM
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
- StemSpan™ Serum-Free Expansion Medium (SFEM) has been developed and tested for the in vitro culture and expansion of human hematopoietic cells, when the appropriate growth factors and supplements are added
- This allows users the flexibility to prepare medium that meets their requirements
- When combined with the appropriate cytokines, SFEM has been used for the culture and expansion of hematopoietic cells isolated from other species, including mouse, non-human primate, and dog
- SFEM has also been used for culture of various other hematopoietic and non-hematopoietic cell types
- Using appropriate StemSpan™ Expansion Supplements, SFEM may be used to expand CD34+ cells isolated from human cord blood, mobilized peripheral blood, or bone marrow samples, or to expand and differentiate lineage-committed progenitors to generate populations of erythroid, myeloid, or megakaryocyte progenitor cells
- StemSpan™ SFEM II (Catalog #09605) is an improved version of StemSpan™ SFEM that is further enriched to promote and support higher rates of CD34+ expansion and/or cell differentiation
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Expansion |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction