missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies StemSpan™ T Cell Generation Kit

Product Code. 17179961
Encompass
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17179961

Brand: STEMCELL Technologies 09940

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For expansion and differentiation of human CD34+ hematopoietic progenitor cells to T cells

  • Use this two-step protocol to reproducibly expand thousands of CD4+CD8+ double-positive (DP) T cells in cultures initiated with CD34+ human cord blood (CB) cells
  • In step one, CD34+ CB cells are cultured for 14 days in medium containing expansion supplement to promote their proliferation and differentiation into CD7+CD5+ pro-T cells
  • In step two, pro-T cells generated during those first 14 days are cultured for another 4 weeks in medium containing maturation supplement to promote their maturation into DP cells
  • This product is serum- and feeder-free, which helps eliminate variation introduced by serum and stromal cell lines
  • An optional protocol is available to further mature DP cells to CD8 SP T cells
  • This extended protocol uses complete T cell progenitor maturation medium made with reagents included in the StemSpan™ T Cell Generation Kit and which must be combined with ImmunoCult™ CD3/CD28/CD2 T Cell Activator (Catalog #10970 or #10971) and IL-15 (Catalog #78031)

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, T Cells
For Use With (Application) Cell Culture; Differentiation; Expansion
Quantity 1 Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.