Learn More
Greiner Bio-One Sterile Inoculation Loop
Product Code. 10342651
Click to view available options
Color:
Blue
White
Yellow
Quantity:
2000/Pk.
600/Pk.
Unit Size:
Pack of 2000
Pack of 600
Product Code. 10342651
Brand: Greiner Bio-One 731181
Currently Unavailable: We have no information when this product will be available.
View alternative products
Transfers inoculum for streaking. Greiner Bio-One™ Sterile Inoculation Loops are ideal for retrieving an inoculum from a culture of microorganisms.
  • Flexible loops for easier collection and inoculation
  • Sterilized by irradiation
  • For research only; not for use in diagnostic procedures, unless otherwise indicated
TRUSTED_SUSTAINABILITY
Specifications
Disposable Single-use
Length (Metric) 200 mm
Sterility Sterile
Color Yellow
Quantity 600/Pk.
For Use With (Application) Research use