Learn More
Hartmann Sterillium Gel Pure Hand Disinfectant
Click to view available options
Volume (Metric):
475 mL
Container Type:
Dispenser Compatible
Unit Size:
475mL
Description
Sterillium Gel pure is suitable for hygienic and surgical hand disinfection for particularly sensitive skin.
- Based on ethanol
- Efficacy: bactericidal, levurocidal, fungicidal, tuberculocidal, mycobactericidal, limited virucidal (incl. HBV, HIV, HCV), limited virucidal PLUS, virucidal
- Norovirus efficacy within hygienic hand disinfection (tested on murine norovirus)
- Limited virucidal PLUS
- Increases skin moisture with regular use
- Colorant and perfume-free
- Leaves no sticky residues
Specifications
Specifications
| Product Type | Disinfectant |
| Scent | Unscented |
| Appearance | Colorless |
| Boiling Point | 17.1°C |
| Composition | Includes ethanol |
| Container Type | Dispenser Compatible |
| Volume (Metric) | 475 mL |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P501-Dispose of contents/container to .