Learn More
Sampling Systems™ SteriWare™ Disposable Knife
BIOPHARMA
Product Code. 15859421 Shop All Sampling Systems Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Product Code. 15859421
Brand: Sampling Systems™ 8039A100
Only null left
Add to basket
Add to basket
Use straight from the bag without the need for costly cleaning. Sampling Systems™ SteriWare™ Disposable Knife is ideal for preparing samples as well as general laboratory use.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No Cross Contamination
  • No Cleaning required
Specifications
Sterility Non-sterile
Color White
Disposable Single-use
Length (Metric) Handle 100 mm
Material HDPE
Length (Metric) 160 mm