Learn More
CEEC Controlled Environment Equipment Corporation™ Sticky Mats
Product Code. p-8010419
Click to view available options
Color:
Blue
White
Dimensions (L x W):
45 x 91 cm
46 x 91cm
61 x 91 cm
66 x 91 cm
45 x 114 cm
66 x 114 cm
Unit Size:
Pack of 120
Product Code. 11866373
Brand: CEEC Controlled Environment Equipment Corporation™ CSS243634TWW
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Eliminate dust contamination from soles with CEEC Controlled Environment Equipment Corp.™ Sticky Mats. They adhere directly to a floor or in an aluminum frame for decontaminating shoes upon entering a cleanroom environment.
  • Adhesive dust mats
  • Sheets numbered from 30 to 1
  • To stick directly to the floor or in an aluminum frame
Specifications
Dimensions (L x W) 61 x 91 cm
Quantity 120/Pk.
Color White