missing translation for 'onlineSavingsMsg'
Learn More

Kartell Stirring Bar

Product Code. 18007761
Click to view available options
Length (Metric):
12 mm
25 mm
30 mm
40 mm
50 mm
64 mm
Diameter (Metric):
3 mm
6 mm
8 mm
16 mm
20 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 18007761

Brand: Kartell 0068500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Magnetic PTFE coated stirring bar suitable fo food stuff.

  • PTFEs (Polytetrafluoroethylene) are polymers with high molecular weight that are widely used owing to their optimum chemical, electrical and thermal properties; their friction coefficient is very low
  • They can be used in the more corrosive environments and do not contaminate the more sensitive materials in ultra-pure or corrosive applications
  • Extremely high electrical resistance low dielectric constant and low dielectric loss factor
  • Self-lubricating and shockproof features, toughness and flexibility even at low temperatures
  • Very high chemical resistance to acids, alkali and solvents is typical of the fluoride polymers; it is therefore used for components in the petrochemical and chemical industry alimentary: physiologically inert, it is approved by some Authorities for uses in contact with foods electrical: excellent dielectric, self-extinguishing and stability in bad weather features make it increasingly used in this sector mechanical: its low friction coefficient makes it usable for very low load applications
  • The magnetic stir bars shown on the following pages have internal magnets in Alnico V that gives them of high duration and power features
  • These magnets are encapsulated in first quality PTFE cases
  • The strict quality controls applied to these products ensure correct positioning of the magnetic core, the quality of the external surface, breakage resistance and constancy of the thickness of the external covering.

Specifications

Autoclavable Autoclavable
Coating PTFE
Diameter (Metric) 16 mm
Length (Metric) 30 mm
Material PTFE
Quantity 10/Pk.
Shape Oval
Type Magnetic Stir Bar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.