Learn More
Fisherbrand™ Stainless Steel Stirring Shafts with Propellers
Product Code. 15113007 Shop All Fisherbrand Products
Click to view available options
Size:
305mm
457mm
610mm
762mm
Includes:
Propellor, 108mm
Propellor, 63.5mm
Unit Size:
Each
Product Code. 15113007
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Excellent at producing axial flows in vessels from top to bottom. Fisherbrand™ Stainless Steel Stirring Shafts with Propellers are for use with overhead stirrer units.
  • Available in a range of sizes
  • Resistant to corrosion
Specifications
Includes Propellor, 63.5mm
Quantity 1 Each
Size 457mm
Type Stirring Shaft