Learn More
Kartell™ Stopper, conical, red rubber, upper\lower diameters 16 x 12mm, height 20mm
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
Brand:  Kartell™ 0113400
Additional Details : Weight : 0.01000kg
Product Code. 12647805
  • 11.25€ / Pack of 25
Estimated Shipment: 17-05-2024
to see stock.
Description
Description
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
  • For general purpose use
  • Red rubber
  • Conical shape
  • Size: lower (bottom) diameter x height
Specifications
Specifications
Red
20mm
12mm
16mm
Rubber
General purpose laboratory
12 x 20mm
Conical Stopper
25/Pk.
Conical