Learn More
Kartell™ Stopper, conical, red rubber, upper\lower diameters 37 x 28mm, height 37mm
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
Brand:  Kartell™ 0114200
Additional Details : Weight : 0.01000kg
Product Code. 12657805
  • 19.20€ / Pack of 5
Estimated Shipment: 19-04-2024
to see stock.
Description
Description
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
  • For general purpose use
  • Red rubber
  • Conical shape
  • Size: lower (bottom) diameter x height
Specifications
Specifications
Red
37mm
28mm
25/Pk.
Conical
General purpose laboratory
28 x 37mm
37mm
Rubber