Learn More
Kartell™ Red Rubber Conical Stoppers
Kartell™ Red Rubber Conical Stoppers are suitable for high temperatures and are used in combination with a flask, bottle, or tube.
Brand:  Kartell™ 0114500
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12631026
  • 31.14€ / Pack of 5
Estimated Shipment
03-06-2021
Add to basket
Description
Description
  • For general purpose use
  • Red rubber
  • Conical shape
  • Size: lower (bottom) diameter x height
Specifications
Specifications
General purpose laboratory
25/Pk.
Red
48mm
Conical Stopper
50mm
37 x 50mm
37mm
Rubber
Conical