Learn More
Masterflex™ Straight Connector
Click to view available options
:
For 3/32 x 1/8 in. tubing I.D.
Specifications
Specifications
| Overall Length (English) | 4.8 in. |
| For Use With | 3/32 x 1/8 in. tubing I.D. |
| Packaging Quantity | 100 Pack |
| Tubing Size | 3/32 in |
| Hose Barb | Barbed |
| Width (English) | 4.7 in. |
| Shape | Straight |
| Height (English) | 0.7 in. |