Learn More
DWK Life Sciences Wheaton™ Straight-sided Glass Jar
Product Code. p-7111684 Shop All Wheaton Science Products Products
Click to view available options
Closure Liner:
Polyvinyl
No. per Case:
48/Case
missing translation for 'unitSize'
Pack of 48
Product Code. 15571980
missing translation for 'mfr': DWK Life Sciences W217906
Only null left
Add to basket
Add to basket
Straight-sided wide-mouth bottles with white polypropylene screw caps. Made of Type III clear soda lime glass.
  • Straight smooth walls allow for complete removal of contents
  • Ideal for soil sampling and environmental applications
  • With attached screw caps lined in PTFE or PV
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 30 mL
Color Clear
Material Soda Lime Glass (Type 3)
No. per Case 48/Case
Closure Color White
Mouth Wide
Product Type Straight-sided Jar
Closure Liner Polyvinyl
Closure Material Polypropylene