Learn More
DWK Life Sciences Wheaton™ Straight-sided Glass Jar
Shop All Wheaton Science Products Products
Click to view available options
Closure Liner:
Polyvinyl
No. per Case:
48/Case
missing translation for 'unitSize'
Pack of 48
Description
- Straight smooth walls allow for complete removal of contents
- Ideal for soil sampling and environmental applications
- With attached screw caps lined in PTFE or PV
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Color | Clear |
| Material | Soda Lime Glass (Type 3) |
| No. per Case | 48/Case |
| Closure Color | White |
| Mouth | Wide |
| Product Type | Straight-sided Jar |
| Closure Liner | Polyvinyl |
| Closure Material | Polypropylene |