Learn More
IBA Lifesciences GmbH Strep-Tactin™ APC for Fab Streptamers™
Product Code. 16431485 Shop All IBA Lifesciences Products
Click to view available options
:
Includes: All required reagents for cell isolation
Product Code. 16431485
Brand: IBA Lifesciences GmbH 65011005
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
IBA Lifesciences GmbH Strep-Tactin™ APC for Fab Streptamers™ is for reversible cell isolation delivering label-free target cells.
  • Low-affinity Fab-Strep molecules are reversibly multimerized on Strep-Tactin™ PE or APC forming a fluorescent Fab Streptamer™ for cell staining
  • Treatment of labeled target cells with the competing Strep-Tactin™ ligand biotin causes a disruption of the Fab Streptamer™ complex and results in spontaneous dissociation of the Fab-Streps from the target cells
  • The result is a functional and label-free target cell
Specifications
Includes All required reagents for cell isolation
For Use With (Application) For 5 x 107 to 1 x 108 cells