Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ High Capacity Resin
Product Code. 15709438 Shop All IBA Lifesciences Products
Change view
Click to view available options
Capacity (Metric):
4 mL
20 mL
50 mL
Unit Size:
20mL
4mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15709438 50 mL 50mL
15729438 20 mL 20mL
15868371 4 mL 4mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15709438 Supplier IBA Lifesciences Supplier No. 24030025
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Strep-Tactin™XT Superflow™ high capacity resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT Superflow™ high capacity is a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • It allows the addition of denaturing agents such as urea during the purification procedure. In comparison to Strep-Tactin™ XT Superflow™, Strep-Tactin™XT Superflow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 3- to 5-fold higher yield of purified protein.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary. 
Specifications
Capacity (Metric) 50 mL
Type High Capacity Resin