missing translation for 'onlineSavingsMsg'
Learn More

Cytiva StrepTactin™ Sepharose™ High Performance Medium

Mild purification conditions preserve antigenicity and function. Cytiva StrepTactin Sepharose High Performance media is used for robust, high resolution purification of Strep II-tagged proteins.

Brand:  Cytiva 28-9356-00

Additional Details : CAS : 79-06-1 Weight : 0.11400kg


 View more versions of this product

Product Code. 10762827

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • Medium with high binding capacity yields high resolution, sharp concentrated peaks, and reproducible purifications
  • Physiological conditions and mild elution preserves the activity of the target protein
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Easily scaled up, and easily regenerated with 0.5M NaOH
  • Convenient use with a peristaltic pump or chromatography system

  • Matrix: rigid, highly cross-linked 6% agarose
  • Ave. particle size: 34μm
  • Dynamic binding capacity (protein dependent): approx. 6mg Strep(II)-tagged protein/mL medium
  • Compatibility: stable in all commonly used buffers, 0.5M NaOH (regeneration and cleaning), reducing agents, denaturants, and detergents
  • pH stability: >7

Specifications

Specifications

StrepTactin Sepharose HP media
<300 cm/hr.
7
6% Highly Cross-linked Agarose
Purification of Strep (II) -tagged recombinant Proteins to high purities in concentrated forms and small volumes by using with perstaltic pump or chromato graphy system
50mL
StrepTactin (∼5mg StrepTactin/mL medium)
34μm
2° to 8°C
Strep-Tactin Sepharose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • StrepTactin Sepharose High Performance, 50 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Promotions

Promotions