missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Streptamer™ Solution Set XL

Streptamer Solution Set XL is designed to be used for washing and dissociation.

Brand:  IBA Lifesciences 6-5603-055

Additional Details : Weight : 0.55500kg

Product Code. 15683888

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • Includes 11 x 50mL Streptamer™ washing buffer (Buffer IS, 10x)
  • Includes 5 x 1mL D-Biotin stock solution for Streptamer™ Technology
  • Both solutions are sterile filtered and do not contain any preservatives
  • Please handle under sterile conditions
Specifications

Specifications

11 x 50 mL Washing buffer, 5 x 1 mL Stock Solution
Washing buffer and D-Biotin stock solution
Washing and Dissociation of Streptamer™ Reagents From Cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions