missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 450 Conjugate
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Specifications
Specifications
| Concentration | 0.2 mg/mL |
| Conjugate | eFluor 450 |
| Quantity | 100 μg |
| Form | Liquid |
| Product Line | eBioscience |
| Product Type | Streptavidin Conjugate (Fluorescent) |
| Target Molecule | Biotin |
| Label or Dye | eFluor 450 |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction