Learn More
Invitrogen™ eBioscience™ Streptavidin PerCP-Cyanine5.5 Conjugate
Product Code. 15311180
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
Contains Sodium azide
Product Code. 15311180
Brand: Invitrogen™ 45431780
Contains Sodium azide
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 0.2 mg/mL
Conjugate PerCP-Cy5.5
Quantity 25 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
Label or Dye PerCP-Cy5.5