missing translation for 'onlineSavingsMsg'
Learn More

Melford Streptomycin Sulfate

Product Code. p-200087301
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
This item is not returnable. View return policy

Product Code. 30018816

Brand: Melford S6200050.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Streptomycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, disrupting the initiation complex and leading to misreading of mRNA.

Streptomycin Sulfate is an aminoglycoside antibiotic that inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit, disrupting the initiation complex and leading to misreading of mRNA. This interference prevents proper translation and inhibits bacterial growth.

Streptomycin is employed in microbiology for selective bacterial isolation and antibiotic sensitivity testing to assess the susceptibility of bacterial strains. Ideal for studying bacterial susceptibility patterns and antibiotic resistance. Its inclusion in culture media aids in selectively isolating bacteria susceptible to this antibiotic.

Specifications

Chemical Name or Material Streptomycin Sulfate
CAS 3810-74-0
Quantity 50 g
Molecular Formula C42H78N14O24-3H2SO4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.